Anti-BCAS2 Rabbit Polyclonal Antibody
Catalog # 10450-948
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Breast carcinoma-amplified sequence 2
- Antigen Symbol:BCAS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:10286
- Antigen Synonyms:Breast carcinoma-amplified sequence 2|BCAS2|Pre-mRNA-splicing factor SPF27|SPF27|DAM1|Snt309
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:162-210/225
- Purification:Purified by Protein A
- Cat. No.:10450-948
- Supplier no.:BS-6610R
Specifications
About this item
Component of the PRP19-CDC5L complex that forms an integral part of the spliceosome and is required for activating pre-mRNA splicing. May have a scaffolding role in the spliceosome assembly as it contacts all other components of the core complex.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: BCAS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat