Anti-ADAMTS10 Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10450-478
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:ADAM Metallopeptidase With Thrombospondin Type 1 Motif, 10
- Antigen Symbol:ADAMTS10
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:81794
- Antigen Synonyms:WMS|ADAM metallopeptidase with thrombospondin type 1 mot 10|A disintegrin and metalloproteinase with thrombospondin mots 10|EC 3.4.24|ADAM TS10|A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 mot|ATS10_HUMAN.|10
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10450-478
- Supplier no.:BS-6588R-CY3
Specifications
About this item
ADAMTS10 is a member of the ADAMs family of proteinases with Thrombospondin motifs. The catalytic site of ADAMTS10 is typical of the metalloproteinase catalytic domains, with an HExxHxxxxxH sequence, perhaps giving these enzymes some shared specificity. ADAMTS10 is closest in homology to ADAMTS6, sharing 53% overall identity. Functional mutations in ADAMTS10 have been linked to Weill Marchesani syndrome, a connective tissue disorder marked by fibrillin 1 misprocessing. ADAMTS10 has also been reported to be over expressed in breast cancer tissues and cell lines.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADAMTS10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat