Anti-PLCB2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10447-680
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Phospholipase C Beta 2
- Antigen Symbol:PLCB2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:5330
- Antigen Synonyms:PLC-beta-2|PLCB3|PLC-beta-3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10447-680
- Supplier no.:BS-6472R-FITC
Specifications
About this item
The production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PLCB2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat