Anti-HNRNPC Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10447-460
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Heterogeneous Nuclear Ribonucleoprotein C
- Antigen Symbol:HNRNPC
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:3183
- Antigen Synonyms:C1|HNRNP|SNRPC|HNRPC|C2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10447-460
- Supplier no.:BS-6459R-CY5
Specifications
About this item
Binds pre-mRNA and nucleates the assembly of 40S hnRNP particles. Single HNRNPC tetramers bind 230-240 nucleotides. Trimers of HNRNPC tetramers bind 700 nucleotides. May play a role in the early steps of spliceosome assembly and pre-mRNA splicing. Interacts with poly-U tracts in the 3'-UTR or 5'-UTR of mRNA and modulates the stability and the level of translation of bound mRNA molecules.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HNRNPC
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat