Anti-pan methyl Lysine Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10435-730
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:pan methyl Lysine
- Antigen Symbol:pan methyl Lysine
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10435-730
- Supplier no.:BS-6350R-FITC
Specifications
About this item
Lysine methylation occurs in three distinct states, having either one (me1), two (me2) or three (me3) methyl groups attached to the amine group of the lysine side chain. In eukaryotes, histone H3 trimethylated at lysine 4 (H3K4me3) is associated with active chromatin and gene expression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: pan methyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse