- Antibody Type:Primary
- Antigen Name:Nuclear receptor-interacting protein 1
- Antigen Symbol:NRIP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:8204
- Antigen Synonyms:DCAF6_HUMAN.|IQWD1v MSTP055|NRIP1|ARCAP|IQ mot and WD repeats 1|Androgen receptor complex associated protein|Nuclear receptor interaction protein|RP4 745I14.1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10432-928
- Supplier no.:BS-6210R-A350
WD-repeat proteins are a large family of eukaryotic proteins coordinating multi-protein complex assemblies. Their role has been implicated in multiple cellular processes including signal transduction, transcriptional regulation, cell cycle control and apoptosis. NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up-regulates their transcriptional activity, thereby functioning as a nuclear receptor co-activator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer, thus indicating a potential therapeutic intervention. Northern Blot analysis detects a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NRIP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat