Anti-RASAL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10430-280
: Bioss
- Antibody Type:Primary
- Antigen Name:RAS Protein Activator Like 1
- Antigen Symbol:RASAL
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:8437
- Antigen Synonyms:RAS protein activator like 1 (GAP1 like)|GAP1 like protein|RASAL|RAS-GTPase-activating protein-like|RAS protein activator like 1|RASAL1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:460-510/805
- Purification:Purified by Protein A
- Cat. No.:10430-280
- Supplier no.:BS-6088R-FITC
Probable inhibitory regulator of the Ras-cyclic AMP pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RASAL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat