Anti-RASA3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10427-106
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Ras GTPase-activating Protein 3
- Antigen Symbol:RASA3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:22821
- Antigen Synonyms:GAPIII|GAP1IP4BP
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:510-560/834
- Purification:Purified by Protein A
- Cat. No.:10427-106
- Supplier no.:BS-5954R-FITC
Specifications
About this item
Inhibitory regulator of the Ras-cyclic AMP pathway. Binds inositol tetrakisphosphate (IP4) with high affinity. Might be a specific IP4 receptor.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RASA3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat