Anti-PRODH Rabbit Polyclonal Antibody (Cy5®)
10424-184
: Bioss
- Antibody Type:Primary
- Antigen Name:Proline Dehydrogenase (oxidase) 1
- Antigen Symbol:PRODH
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:5625
- Antigen Synonyms:PIG6|POX|HSPOX2|TP53I6|PRODH1|PRODH2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:170-220/600
- Purification:Purified by Protein A
- Cat. No.:10424-184
- Supplier no.:BS-5813R-CY5
Proline oxidase catalyzes the conversion of proline to pyrroline-5-carboxylate, or P5C during the degradation of the amino acid Proline. Defects in PRODH are the cause of hyperprolinemia type 1, a disorder characterized by elevated serum proline levels. Defective PRODH may be involved in the psychiatric and behavioral phenotypes associated with the 22q11 velocardiofacial and DiGeorge syndrome and may be associated with susceptibility to schizophrenia 4 (SCZD4).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PRODH
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat