Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-DSTYK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-DSTYK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10423-990
 :  Bioss
Anti-DSTYK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10423-990
 :  Bioss
 :  BS-5805R-FITC

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Dual serine/threonine And tyrosine protein kinase
  • Antigen Symbol:
    DSTYK
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8737
  • Antigen Synonyms:
    SGK496|Receptor interacting protein kinase 5|RIP homologous kinase|HDCMD38P|RIP5|KIAA0472|DUSTY_HUMAN.|Sugen kinase 496|Dusty protein kinase|Receptor interacting serine/threonine protein kinase 5|Dusty PK|Receptor interacting protein 5
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    600-650/671
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10423-990
  • Supplier no.:
    BS-5805R-FITC

 

 

Serine-threonine kinase which transduces inflammatory and cell-death signals (programmed necrosis) following death receptors ligation, activation of pathogen recognition receptors (PRRs), and DNA damage. Upon activation of TNFR1 by the TNF-alpha family cytokines, TRADD and TRAF2 are recruited to the receptor. Phosphorylates DAB2IP at 'Ser-728' in a TNF-alpha-dependent manner, and thereby activates the MAP3K5-JNK apoptotic cascade. Ubiquitination by TRAF2 via 'Lys-63'-link chains acts as a critical enhancer of communication with downstream signal transducers in the mitogen-activated protein kinase pathway and the NF-kappa-B pathway, which in turn mediate downstream events including the activation of genes encoding inflammatory molecules. Polyubiquitinated protein binds to IKBKG/NEMO, the regulatory subunit of the IKK complex, a critical event for NF-kappa-B activation. Interaction with other cellular RHIM-containing adapters initiates gene activation and cell death. RIPK1 and RIPK3 association, in particular, forms a necrosis-inducing complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DSTYK
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat