To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Heat Shock Transcription Factor 1
- Antigen Symbol:HSF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:3297
- Antigen Synonyms:HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- Modification:phospho-Ser303
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10414-080
- Supplier no.:BS-5363R
Specifications
About this item
Two HSFs have been identified in human cells, HSF 1 and HSF 2, which bind to the same HSEs and have 38% sequence identity. These factors are activated by distinct stimuli, HSF 1 is responsive to classical stress signals such as heat, heavy metals and oxidative reagents, whereas HSF 2 is activated during hemin-mediated differentiation of human erythroleukemia cells.HSF 1 exists constitutively in the cytoplasm and the nucleus of unstressed cells as a monomer which lacks DNA binding activity. Through an unknown signal generated during stress, HSF 1 becomes activated to a nuclear localized, trimeric state which binds to DNA. The phosphorylation of HSF 1 is necessary for maximal transcription of heat shock genes.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat