Anti-CFL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10411-898
: Bioss
- Antibody Type:Primary
- Antigen Name:Cofilin 1 (non-muscle)
- Antigen Symbol:CFL1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:1072
- Antigen Synonyms:HEL-S-15|cofilin|CFL
- Modification:phospho-Tyr140
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:135-142/166
- Purification:Purified by Protein A
- Cat. No.:10411-898
- Supplier no.:BS-5258R-FITC
Binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CFL1
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: