Anti-Chondroitin Sulfate Rabbit Polyclonal Antibody
Catalog # 10410-246
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Chondroitin Sulfate
- Antigen Symbol:Chondroitin Sulfate
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10410-246
- Supplier no.:BS-5178R
Specifications
About this item
Chondroitin sulphate proteoglycans (CSPGs) are upregulated in the central nervous system after injury, specifically around the lesion site where the glial scar forms. CSPGs have been shown to be closely involved in this neuronal growth inhibition, specifically through their sugar chains.Chondroitin sulphate proteoglycans may also play a role in the repair of spinal cord injury.
Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: Chondroitin Sulfate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat