Anti-HEXIM2 Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10408-532
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:hexamethylene bis-acetamide inducible 2
- Antigen Symbol:HEXIM2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:124790
- Antigen Synonyms:L3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10408-532
- Supplier no.:BS-5104R-CY5
Specifications
About this item
Degrades bioactive fatty acid amides like oleamide, the endogenous cannabinoid, anandamide and myristic amide to their corresponding acids, thereby serving to terminate the signaling functions of these molecules. Hydrolyzes polyunsaturated substrate anandamide preferentially as compared to monounsaturated substrates.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HEXIM2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat