Anti-AGPAT2 Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10407-040
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:1-acyl-sn-glycerol-3-phosphate acyltransferase beta
- Antigen Symbol:AGPAT2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:10555
- Antigen Synonyms:LPAAB|BSCL|1-AGPAT2|LPAAT-beta|BSCL1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10407-040
- Supplier no.:BS-5032R-CY5
Specifications
About this item
Agpat2 is a member of the 1-acylglycerol-3-phosphate O-acyltransferase family. It is located within the endoplasmic reticulum membrane and converts lysophosphatidic acid to phosphatidic acid, the second step in de novo phospholipid biosynthesis. Mutations in its have been associated with congenital generalized lipodystrophy (CGL), or Berardinelli-Seip syndrome, a disease characterized by a near absence of adipose tissue and severe insulin resistance. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AGPAT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat