Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CXCL12 Rabbit Polyclonal Antibody (Cy3®)
Anti-CXCL12 Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10405-070
Supplier:  Bioss
Anti-CXCL12 Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10405-070
Supplier:  Bioss
Supplier Number:  BS-4938R-CY3

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Chemokine (C-X-C Motif) Ligand 12
  • Antigen Symbol:
    CXCL12
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    6387
  • Antigen Synonyms:
    SDF1B|TPAR1|SDF1|IRH|SDF1A|Stromal cell-derived factor 1|PBSF|SDF-1|TLSF|SCYB12
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    55-90/93
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10405-070
  • Supplier no.:
    BS-4938R-CY3

Specifications

About this item

Chemoattractant active on T-lymphocytes, monocytes, but not neutrophils. Activates the C-X-C chemokine receptor CXCR4 to induce a rapid and transient rise in the level of intracellular calcium ions and chemotaxis. Also binds to atypical chemokine receptor ACKR3, which activates the beta-arrestin pathway and acts as a scavenger receptor for SDF-1. SDF-1-beta(3-72) and SDF-1-alpha(3-67) show a reduced chemotactic activity. Binding to cell surface proteoglycans seems to inhibit formation of SDF-1-alpha(3-67) and thus to preserve activity on local sites. Acts as a positive regulator of monocyte migration and a negative regulator of monocyte adhesion via the LYN kinase. Stimulates migration of monocytes and T-lymphocytes through its receptors, CXCR4 and ACKR3, and decreases monocyte adherence to surfaces coated with ICAM-1, a ligand for beta-2 integrins. SDF1A/CXCR4 signaling axis inhibits beta-2 integrin LFA-1 mediated adhesion of monocytes to ICAM-1 through LYN kinase. Inhibits CXCR4-mediated infection by T-cell line-adapted HIV-1. Plays a protective role after myocardial infarction. Induces down-regulation and internalization of ACKR3 expressed in various cells. Has several critical functions during embryonic development; required for B-cell lymphopoiesis, myelopoiesis in bone marrow and heart ventricular septum formation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCL12
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: