Anti-GCDFP15 Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10405-688
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Gross Cystic Disease Fluid Protein 15
- Antigen Symbol:GCDFP15
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:GCDGP-15|SABP|Gross cystic disease fluid protein 15|Gp17|GPIP4|Prolactin induced protein|Prolactin inducible protein precursor|Secretory actin binding protein.|GCDGP 15
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10405-688
- Supplier no.:BS-4726R-CY5
Specifications
About this item
Gross cystic disease is a common premenopausal disorder in which gross cysts are the predominant pathologic lesion. It is characterized by production of a fluid secretion which accumulates in the breast cysts. Gross cystic disease fluid is a pathologic secretion from breast composed of several glycoproteins, including a unique 15 kDa monomer protein, GCDFP 15. The cells within the body that produce GCDFP 15 appear to be restricted primarily to those with apocrine function. Studies have found GCDFP 15 to be a highly specific and sensitive marker for breast cancer.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GCDFP15
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat