Anti-TUBB3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10402-898
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Tubulin Beta 3
- Antigen Symbol:TUBB3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:10381
- Antigen Synonyms:Tubulin beta-3 chain|CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:400-450/450
- Purification:Purified by Protein A
- Cat. No.:10402-898
- Supplier no.:BS-4512R-FITC
Specifications
About this item
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain. TUBB3 plays a critical role in proper axon guidance and mantainance.
Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200
Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat