Anti-AURKAIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # 10397-568
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Aurora Kinase AIP1
- Antigen Symbol:AURKAIP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:54998
- Antigen Synonyms:AIP|MRP-S38|AKIP
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:150-199/199
- Purification:Purified by Protein A
- Cat. No.:10397-568
- Supplier no.:BS-4287R-A555
Specifications
About this item
May act as a negative regulator of Aurora-A kinase, by down-regulation through proteasome-dependent degradation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AURKAIP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat