Anti-ABAT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10396-862
: Bioss
- Antibody Type:Primary
- Antigen Name:Aminobutyrate Aminotransferase
- Antigen Symbol:ABAT
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:18
- Antigen Synonyms:GABA-AT|NPD009|GABAT
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:160-210/500
- Purification:Purified by Protein A
- Cat. No.:10396-862
- Supplier no.:BS-4234R-FITC
Catalyzes the conversion of gamma-aminobutyrate and L-beta-aminoisobutyrate to succinate semialdehyde and methylmalonate semialdehyde, respectively. Can also convert delta-aminovalerate and beta-alanine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABAT
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat