Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PRKACG Rabbit Polyclonal Antibody (Cy3®)
Anti-PRKACG Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10390-814
Supplier:  Bioss
Anti-PRKACG Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10390-814
Supplier:  Bioss
Supplier Number:  BS-3964R-CY3

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Protein Kinase cAMP-activated Catalytic Subunit Gamma
  • Antigen Symbol:
    PRKACG
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5568
  • Antigen Synonyms:
    KAPG|PRKACG|PKA C gamma|KAPCG_HUMAN.|Protein kinase cAMP dependent catalytic gamma|Serine threonine protein kinase
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10390-814
  • Supplier no.:
    BS-3964R-CY3

Specifications

About this item

PKA (or cAPK) is a cyclic AMP dependent protein kinase. When activated by the second messenger cAMP, PKA mediates diverse cellular mechanisms, including proliferation, ion transport, regulation of metabolism, plus gene transcription. PKA is comprised of two dimers of two subunits, R (regulatory) and C (catalytic). Two families of R subunit (RI and RII) and three C subunit isoforms (C alpha, C beta, and C gamma) have been identified each possessing distinct cAMP binding properties and resulting in different phosphorylation states. C subunit is activated through autophosphorylation and direct phosphorylation at Thr197 by PDK-1. Tissue specific expression of C gamma, indicates pressure on C gamma during evolution, acting to modulate it in a functionally specific way. Certain amino acid substitutions make C gamma a distinct member of the cAMP dependent subfamily of protein kinases, and suggest that C gamma may be distinct in its protein substrate specificity or its interaction with the different regulatory subunits.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRKACG
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat