Anti-GNB2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10390-396
: Bioss
- Antibody Type:Primary
- Antigen Name:Guanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 2
- Antigen Symbol:GNB2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:2783
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10390-396
- Supplier no.:BS-3942R-FITC
GNB2 belongs to the WD repeat G protein beta family. Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: G protein beta 2/GNB2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat