Anti-GNB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10390-372
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Guanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 1
- Antigen Symbol:GNB1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:2782
- Antigen Synonyms:Transducin beta
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10390-372
- Supplier no.:BS-3941R-FITC
Specifications
About this item
Heterotrimeric guanine nucleotide binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. GNB1 is a beta subunit. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: G protein beta 1/GNB1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat