Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-UCHL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-UCHL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10388-084
 :  Bioss
Anti-UCHL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  10388-084
 :  Bioss
 :  BS-3806R-FITC

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Ubiquitin Carboxyl-Terminal Esterase L1
  • Antigen Symbol:
    UCHL1
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7345
  • Antigen Synonyms:
    Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    30-80/223
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10388-084
  • Supplier no.:
    BS-3806R-FITC

 

 

Ubiquitin-protein hydrolase involved both in the processing of ubiquitin precursors and of ubiquitinated proteins. This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. Also binds to free monoubiquitin and may prevent its degradation in lysosomes. The homodimer may have ATP-independent ubiquitin ligase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat