Anti-LIR6 Rabbit Polyclonal Antibody (Cy3®)
Catalog # 10343-756
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Leukocyte immunoglobulin-like receptor 6
- Antigen Symbol:LIR6
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Leukocyte immunoglobulin-like receptor subfamily A member 1|subfamily A (with TM domain)|member 1 soluble isoform|LILRA1|Leukocyte immunoglobulin-like receptor 6|LIR6|member 1|LIR-6|CD85 antigen-like family member I|LIRA1_HUMAN.|Leukocyte immunoglobulin-like receptor|CD85i
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10343-756
- Supplier no.:BS-2656R-CY3
Specifications
About this item
May act as receptor for class I MHC antigens.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LIR6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human