Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-EIF4E Rabbit Polyclonal Antibody
Anti-EIF4E Rabbit Polyclonal Antibody
Catalog # 10336-972
Supplier:  Bioss
undefined
Anti-EIF4E Rabbit Polyclonal Antibody
Catalog # 10336-972
Supplier:  Bioss
Supplier Number:  BS-2446R

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    eukaryotic translation initiation factor 4E
  • Antigen Symbol:
    EIF4E
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1977
  • Antigen Synonyms:
    eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
  • Modification:
    phospho-Ser209
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    200-217/217
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10336-972
  • Supplier no.:
    BS-2446R

Specifications

About this item

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures. Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit mediates the binding to the mRNA cap.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat