Anti-IL2RB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10333-892
: Bioss
- Antibody Type:Primary
- Antigen Name:Interleukin 2 Receptor Beta
- Antigen Symbol:IL2RB
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:3560
- Antigen Synonyms:IL-2RB|CD122|p75|IL-2R subunit beta|high affinity IL-2 receptor beta subunit|beta|CD122 antigen|P70-75|IL15RB|IL-2 receptor subunit beta|interleukin-2 receptor subunit beta|interleukin 15 receptor|high affinity IL-2 receptor subunit beta
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:501-551/551
- Purification:Purified by Protein A
- Cat. No.:10333-892
- Supplier no.:BS-1959R-FITC
Receptor for interleukin-2. This beta subunit is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL2RB
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat