Anti-MRAS Rabbit Polyclonal Antibody (Cy5®)
10331-652
: Bioss
- Antibody Type:Primary
- Antigen Name:Muscle RAS Oncogene homolog
- Antigen Symbol:MRAS
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:22808
- Antigen Synonyms:M-RAs|RRAS3|R-RAS3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10331-652
- Supplier no.:BS-1882R-CY5
MRas is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. They are oncogenically activated in a significant fraction of tumors. MRas participates in reorganisation of actin cytoskeleton. It is also involved in focal adhesion and can activate MAPK signalling.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MRAS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat