Anti-GZMB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10299-950
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Granzyme B
- Antigen Symbol:GZMB
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:3002
- Antigen Synonyms:CSP-B|Granzyme 2 cytotoxic T-lymphocyte-associated serine esterase 1|CCPI|CTSGL1|CGL1|CGL-1|SECT|CSPB|CTLA1|HLP
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:5-55/247
- Purification:Purified by Protein A
- Cat. No.:10299-950
- Supplier no.:BS-1351R-FITC
Specifications
About this item
This enzyme is necessary for target cell lysis in cell-mediated immune responses. It cleaves after Asp. Seems to be linked to an activation cascade of caspases (aspartate-specific cysteine proteases) responsible for apoptosis execution. Cleaves caspase-3, -7, -9 and 10 to give rise to active enzymes mediating apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: pan Granzyme
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat