Specifications
- Antibody Type:Primary
- Antigen Name:Galactosamine-6 Sulfatase
- Antigen Symbol:GALNS
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:2588
- Antigen Synonyms:GALNAC6S|GAS|GalN6S|MPS4A
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10295-352
- Supplier no.:BS-13269R-CY5
Specifications
About this item
Chondroitinase is a 522 amino acid protein that localizes to the lysosome and functions as an exohydrolase that is essential for the degradation of glycosaminoglycans, keratan sulfate and chondroitin 6-sulfate. Using calcium as a cofactor, Chondroitinase, which exists as a disulfide linked oligomer, catalyzes the hydrolysis of the 6-sulfate group on target substrates. Defects in the gene encoding Chondroitinase are the cause of mucopolysaccharidosis type 4A (MPS4A), an autosomal recessive lysosomal storage disease that is characterized by the intracellular accumulation of keratan sulfate and chondroitin-6-sulfate and is associated with dental anomalies, short stature and, in some cases, death in the second or third decade of life.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GALNS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat