Anti-FXYD6 Rabbit Polyclonal Antibody (Cy3®)
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The mammalian FXYD family maintains Na+ and K+ gradients between the intracellular and extracellular milieus of cells in processes such as renal Na+-reabsorption, muscle contraction and neuronal excitability. FXYDs are single-span membrane proteins that share a 35 amino acid signature domain, beginning with the sequence PFXYD and containing seven invariant and six conserved amino acids. Members of the FXYD family include FXYD1 (PLM, phospholemman), FXYD2 (the g subunit of the Na+/K+-ATPase), FXYD3 (Mat8, mammary tumor protein), FXYD4 (CHIF) and FXYD5 (RIC). FXYD6 is expressed in various epithelial cells bordering the endolymph space and in the auditory neurons. FXYD6 co-localizes with Na+/K+-ATPase in the stria vascularis and can be co-immunoprecipitated with Na+/K+-ATPase. After expression, FXYD6 associates with Na+/K+-ATPase alpha1-beta1 and alpha1-beta2 isozymes, which are preferentially expressed in different regions of the inner ear and also with gastric and non-gastric H+/K+-ATPase.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: FXYD6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-13234R-CY3
Datasheet
Specifications
- Size:100 µL
- Cross Adsorption:No
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Isotype:IgG
- Host:Rabbit
- Concentration:1 μg/μl
- Cat. No.:10292-258
- Flow Cytometry:Yes
- Reactivity:Human|Rat|Mouse
- Shipping Temperature:4°C
- Antibody Type:Primary
- Antigen Symbol:FXYD6
- Gene ID:53826
- ImmunoChemistry:Yes
- Conjugation:Cy3®
- Antigen Synonyms:FXYD domain containing ion transport regulator 6|UNQ521/PRO1056|FXYD6_HUMAN.|Phosphohippolin|FXYD domain containing ion transport regulator 6 precursor
- Antigen Name:FXYD Domain Containing ion transport Regulator 6
- Storage Temperature:Store at 4°C for 12 months
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



