Specifications
- Antibody Type:Primary
- Antigen Name:Serine/Arginine-Rich Splicing Factor 10
- Antigen Symbol:SRSF10
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:10772
- Antigen Synonyms:TASR|SFRS13A|TASR1|SFRS13|PPP1R149|SRrp40|FUSIP1|NSSR|SRp38|TASR2|FUSIP2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10292-158
- Supplier no.:BS-13229R-CY7
Specifications
About this item
FUSIP1 is a member of the Serine/Arginine (SR) family of splicing factors. Members of the SR family all contain one or more RNA recognition motifs (RRM) and an SR-rich domain. SR factors are not only essential for constitutive splicing but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. Expressed in a variety of tissues with low expression in kidney, liver and heart, FUSIP1 localizes to the cytoplasm and nuclear speckles. In its dephosphorylated form (occurring during M phase of the cell cycle), FUSIP1 functions as a potent general repressor of pre-mRNA splicing and can interact with U1 SnRNP 70. In its phosphorylated form, FUSIP1 interacts with Tra-2∫ and, together, they may cooperate in the regulation of splicing. Four isoforms exist for FUSIP1. In neurons, FUSIP1 isoforms may act to either positively or negatively regulate alternative splicing.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SRSF10
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat