Anti-SDC1 Rabbit Polyclonal Antibody (Cy5®)
10292-360
: Bioss
- Antibody Type:Primary
- Antigen Name:Syndecan 1
- Antigen Symbol:SDC1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:6382
- Antigen Synonyms:SDC|Syndecan 1|SDC1|SYND 1|CD 138|SYND1|CD138|Syndecan1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:275-310/310
- Purification:Purified by Protein A
- Cat. No.:10292-360
- Supplier no.:BS-1309R-CY5
Cell surface proteoglycan that bears both heparan sulfate and chondroitin sulfate and that links the cytoskeleton to the interstitial matrix.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SDC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat