To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:dual adaptor of phosphotyrosine And 3-phosphoinositides
- Antigen Symbol:DAPP1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:27071
- Antigen Synonyms:BAM32|b lymphocyte adapter protein Bam32|hDAPP1|DKFZp667E0716|dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide|B-cell adapter molecule of 32 kDa
- Modification:phospho-Tyr139
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10285-600
- Supplier no.:BS-12983R-CY5
Specifications
About this item
B cell adapter molecule is also designated dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide (DAPP1) or B lymphocyte adapter protein. BAM32 is a B cell-associated adapter that is crucial for B cell antigen receptor signaling regulation. BAM32 interacts with Ptdlns and PLC g2 and, upon B cell activation, the protein is phosphorylated on tyrosine residues. It is a mainly cytoplasmic protein that can translocate to the cell membrane after cell stimulation. BAM32, which contains one PH domain and one SH2 domain, is primarily expressed in placenta and lung tissues, but can also be detected in heart, liver, pancreas and brain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DAPP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat