Specifications
- Antibody Type:Primary
- Antigen Name:Catecholamine-sulfating Phenol Sulfotransferase
- Antigen Symbol:CSPS
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:101060714
- Antigen Synonyms:STM|Monoamine-sulfating phenol sulfotransferase|Thermolabile phenol sulfotransferase|Monoamine sulfating phenosulfotransferase|Monoamine preferring sulfotransferase|FLJ54864|Sulfotransferase family cytosolic 1A phenol preferring member 3|TL-PST|TLPST.|HAST3|ST1A3_HUMAN|Sulfotransferase 1A3/1A4|Aryl sulfotransferase|Sulfotransferase family cytosolic 1A phenol preferring member 4|Catecholamine Sulfating Phenol Sulfotransferase|Phenol sulfotransferase 1A5|Catecholamine-sulfating phenol sulfotransferase|MPST|Sulfotransferase|M-PST|HAST|TL PST|Sulfokinase|Placental estrogen sulfotransferase|SULT1A4|Sulfotransferase monoamine preferring|Dopamine specic sulfotransferase|Aryl sulfotransferase 1A3/1A4|SULT1A 3|monoamine-preferring|M PST|ST1A3/ST1A4|Thermolabile monoamine M form phenol sulfotransferase|ST1A5
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10288-602
- Supplier no.:BS-12945R-CY3
Specifications
About this item
Cysteine string proteins (CSPs) are synaptic vesicle-associated, secretory vesicle proteins that are involved in Ca2+-regulated exocytosis of synaptic vesicles and modulation of presynaptic transmembrane calcium fluxes in neuroendocrine and endocrine cell types. CSP contains a J-domain that binds HSP 70/HSC 70 chaperone ATPases and a membrane-targeting, palmitoylated cysteine-rich string region. CSPs may act as molecular chaperones in synapses, and mediate conformational folding of components of the vesicular exocytotic machinery. CSP is involved in the fine tuning of neurotransmission through its interaction with receptor-coupled trimeric GTP binding proteins (G proteins) and N-type Ca2+ channels. Two variants of CSP have been described: CSP1; and the 31 amino acid, C-terminally truncated isoform, CSP2. Subcellular fractionation of insulinoma cells shows CSP1 in granular fractions, while the membrane and cytosol fractions contain predominantly CSP2. The fractions also contain additional proteins, presumably CSP dimers. Furthermore, in various mammalian cell lines (including rat brain) CSP1 expression predominates CSP2 expression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CSPS
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: