Anti-REL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10288-236
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:V-rel Avian Reticuloendotheliosis Viral Oncogene homolog
- Antigen Symbol:REL
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:5966
- Antigen Synonyms:4.1-G|c-Rel|4.1G
- Modification:phospho-Ser503
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10288-236
- Supplier no.:BS-12918R-FITC
Specifications
About this item
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA (MIM 164014), RELB (604758), NFKB1 (MIM 164011), and NFKB2 (MIM 164012). These proteins are related through a highly conserved N-terminal region termed the 'Rel domain,' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (MIM 164008) (Belguise and Sonenshein, 2007 [PubMed 18037997]).[supplied by OMIM, May 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: REL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse