Anti-APOBEC2 Rabbit Polyclonal Antibody (Cy7®)
10282-552
: Bioss
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B mRNA Editing Enzyme, Catalytic Polypeptide-Like 2
- Antigen Symbol:APOBEC2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:10930
- Antigen Synonyms:ARCD1|catalytic polypeptide 2|catalytic polypeptide-like 2 |OTTHUMP00000039773|Apolipoprotein B mRNA editing enzyme|ARP1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10282-552
- Supplier no.:BS-12493R-CY7
APOBEC2 is a 224 amino acid protein that belongs to the cytidine and deoxycytidylate deaminase family. Expressed exclusively in heart and skeletal muscle, APOBEC2 is thought to be a probable C (cytidine) to U (uridine) editing enzyme. However, unlike other members of the family, such as APOBEC1, which mediates the editing of apolipoprotein (apo) B mRNA, APOBEC2 does not display any detectable apoB mRNA editing activity. Also, APOBEC2 has been shown to have low, but definite, intrinsic cytidine deaminase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat