Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-APOBEC2 Rabbit Polyclonal Antibody (Cy5®)
  10282-550
 :  Bioss
Anti-APOBEC2 Rabbit Polyclonal Antibody (Cy5®)
  10282-550
 :  Bioss
 :  BS-12493R-CY5

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Apolipoprotein B mRNA Editing Enzyme, Catalytic Polypeptide-Like 2
  • Antigen Symbol:
    APOBEC2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10930
  • Antigen Synonyms:
    ARCD1|catalytic polypeptide 2|catalytic polypeptide-like 2 |OTTHUMP00000039773|Apolipoprotein B mRNA editing enzyme|ARP1
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10282-550
  • Supplier no.:
    BS-12493R-CY5

 

 

APOBEC2 is a 224 amino acid protein that belongs to the cytidine and deoxycytidylate deaminase family. Expressed exclusively in heart and skeletal muscle, APOBEC2 is thought to be a probable C (cytidine) to U (uridine) editing enzyme. However, unlike other members of the family, such as APOBEC1, which mediates the editing of apolipoprotein (apo) B mRNA, APOBEC2 does not display any detectable apoB mRNA editing activity. Also, APOBEC2 has been shown to have low, but definite, intrinsic cytidine deaminase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat