- Antibody Type:Primary
- Antigen Name:MFNG O-Fucosylpeptide 3-Beta-N-Acetylglucosaminyltransferase
- Antigen Symbol:MFNG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:4242
- Antigen Synonyms:EC 2.4.1.222|Manic Fringe Homolog
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10283-834
- Supplier no.:BS-12390R-FITC
Three mammalian fringe family members, Manic, Radical and Lunatic Fringe, have been identified as proteins related to Drosophila fringe, a protein involved in development. Fringe proteins act upstream of the Notch signaling pathway and are involved in boundary determination during segmentation. Each mammalian fringe displays different patterns of expression, though all are expressed in the mouse embryo as well as in many adult tissues. Radical fringe plays a key role in the development of the limb bud. Lunatic fringe is required for normal somite segmentation and patterning and is thought to be a target of the molecular clock. Manic fringe, also involved in somatic development, has been shown to render mouse NIH/3T3 cells tumorigenic in SCID mice.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MFNG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat