- Antibody Type:Primary
- Antigen Name:Multiple EGF-Like-Domains 10
- Antigen Symbol:MEGF10
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:84466
- Antigen Synonyms:multiple EGF-like-domains 10|Multiple epidermal growth factor-like domains 10|MEG10_HUMAN|Multiple EGF-like domains protein 10|MEGF protein|megf10|Multiple epidermal growth factor-like domains protein 10.
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10283-458
- Supplier no.:BS-12372R-CY5
MEGF10 is a 1,140 amino acid protein encoded by the human gene MEGF10. MEGF10 belongs to the MEGF family and contains fifteen EGF-like domains and one EMI domain. The engulfment of dying cells is a specialized form of phagocytosis that is highly conserved across evolution and is mediated by the concerted action of several molecules. MEGF10 is an engulfment receptor protein that localizes to the plasma membrane in a punctuated pattern. The MEGF10 protein shares structural similarities with nematode engulfment receptor cell death abnormal-1 (CED-1). MEGF10 function is modulated by the ATP binding cassette transporter ABCA1, which is an ortholog to CED-7. It is expressed at the cell surface in clusters around cell corpses during engulfment, and during the engulfment of apoptotic thymocytes is recruited to the bottom of the forming phagocytic cup.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MEGF10
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat