Anti-TCP1A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # 10277-778
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:T-complex Protein 1 Subunit Alpha
- Antigen Symbol:TCP1A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:6950
- Antigen Synonyms:TRic|Tp63|c-cpn|Ccta|Cct1|p63|Tcp-1|CCT|AI528772
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10277-778
- Supplier no.:BS-12292R-A350
Specifications
About this item
Molecular chaperone; assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. Known to play a role, in vitro, in the folding of actin and tubulin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TCP1A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat