Anti-ZNF473 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10276-754
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Zinc Finger Protein 473
- Antigen Symbol:ZNF473
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:25888
- Antigen Synonyms:Zinc finger protein 473|zinc finger protein ZFP100|HZFP100|Zinc finger protein 100 homolog|ZN473_HUMAN|Zfp 100|Zfp-100|ZNF473.
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10276-754
- Supplier no.:BS-12242R-FITC
Specifications
About this item
Involved in histone 3'-end pre-mRNA processing by associating with U7 snRNP and interacting with SLBP/pre-mRNA complex. Increases histone 3'-end pre-mRNA processing but has no effect on U7 snRNP levels, when overexpressed. Required for cell cycle progression from G1 to S phases.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZNF473
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat