Anti-DDX3X Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10275-088
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked
- Antigen Symbol:DDX3X
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:1654
- Antigen Synonyms:DDX14|DBX|DDX3|HLP2|CAP-Rf
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10275-088
- Supplier no.:BS-12189R-FITC
Specifications
About this item
DDX3 is involved in RNA metabolism. Two DDX3 paralogs are found in humans; DDX3X is encoded by a gene found on the X chromosome while DDX3Y is encoded by a gene on the Y chromosome. DDX3Y is exclusively expressed in testis and is required for normal spermatogenesis. DDX3X is ubiquitously expressed and predominantly localizes to the nuclear speckles, participating in RNA splicing, transcription, translation initiation, mRNA transport and cell cycle regulation. DDX3X also partakes in HIV-1 replication and hepatitis C viral infections.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DDX3X
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat