Anti-TP53I5 Rabbit Polyclonal Antibody (Cy5®)
10275-516
: Bioss
- Antibody Type:Primary
- Antigen Name:Tumor Protein 53-inducible Protein 5
- Antigen Symbol:TP53I5
- Clonality:Polyclonal
- Conjugation:Cy5®
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:TMEM16J|PIG5|TP53I5|Ano9|Anoctamin-9|ANO9_HUMAN|p53-induced gene 5 protein|Transmembrane protein 16J|Tumor protein p53-inducible protein 5.
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10275-516
- Supplier no.:BS-12159R-CY5
May act as a calcium-activated chloride channel.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TP53I5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat