To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Pregnancy Up-Regulated Nonubiquitous CaM Kinase
- Antigen Symbol:PNCK
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:BSTK3|CaMK1b
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10274-372
- Supplier no.:BS-12134R-FITC
Specifications
About this item
The Ca2+/calmodulin-dependent protein kinases (CaMKs) comprise a structurally related subfamily of serine/threonine kinases. CaMKI Beta (Ca2+/calmodulin-dependent protein kinase type 1B), also known as PNCK (pregnancy up-regulated non-ubiquitously expressed CaM kinase) or BSTK3, is a 343 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one protein kinase domain. Existing as multiple alternatively spliced isoforms, CaMKI Beta functions to catalyze the ATP-dependent phosphorylation of CaMKI, an event that activates CaMKI activity and may be important for Ca2+-triggered signaling cascades within the cell. The gene encoding CaMKI Beta maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PNCK
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat