Specifications
- Antibody Type:Primary
- Antigen Name:5-hydroxytryptamine receptor 1E
- Antigen Symbol:5HT1E
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Serotonin receptor 1E|S31|5 hydroxytryptamine serotonin receptor 1E|5 hydroxytryptamine 1E receptor|5-hydroxytryptamine receptor 1E|5HT1E_HUMAN.|5 hydroxytryptamine serotonin receptor 1E G protein coupled|5-HT-1E|HTR1E
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10272-324
- Supplier no.:BS-12047R-FITC
Specifications
About this item
The members of the G-protein-coupled receptor family are distinguished by their slow transmitting response to ligand binding. These seven transmembrane proteins include the adrenergic, serotonin and dopamine receptors. The effect of the signaling molecule can be excitatory or inhibitory depending on the type of receptor to which it binds. b-adrenergic bound to adrenaline activates adenylyl cyclase, while a2-adrenergic receptor bound to adrenaline inhibits adenylyl cyclase. Like the a2-adrenergic receptor, serotonin receptor functions are also mediated by G proteins that inhibit the activity of adenylyl cyclase. The serotonin receptors have been classified into several categories, designated SR-1–7 (5HT1–7). Subtypes within the SR-1 group include SR-1A, -1B, -1D, -1E and -1F.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: 5HT1E
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human