To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Leucine Rich Repeat Transmembrane Neuronal 2
- Antigen Symbol:LRRTM2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Leucine-rich repeat neuronal 2 protein|LRRN2|Leucine-rich repeat transmembrane neuronal protein 2|LRRTM2|KIAA0416
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10267-294
- Supplier no.:BS-11877R-CY5
Specifications
About this item
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic Alpha/Beta horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. The LRRTM protein family plays a role in the regulation of various cellular events during nervous system development. Localizing predominantly to the nervous system, LRRTM family members are known to exhibit synaptogenic activity. LRRTM2 (leucine rich repeat transmembrane neuronal 2), also known as LRRN2, is a 516 amino acid single-pass type I membrane protein involved in the development maintenance of the vertebrate nervous system. Expressed in kidney and neuronal tissues, LRRTM2 contains ten LRR repeats and belongs to the LRRTM family. LRRTM2 is encoded by a gene that maps to human chromosome 5q31.2.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: LRRTM2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat