- Antibody Type:Primary
- Antigen Name:AVP Receptor V1b
- Antigen Symbol:AVPR1B
- Clonality:Polyclonal
- Conjugation:Cy7®
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:VPR3.|Arginine vasopressin receptor 1B|V1BR_HUMAN|Vasopressin V1b receptor|AVPR V3|Vasopressin V3 receptor|AVPR V1b|V1bR|Pituitary vasopressin receptor 3|AVPR3|Arginine vasopressin receptor 3|Avpr1b|Antidiuretic hormone receptor 1b
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10262-862
- Supplier no.:BS-11800R-CY7
Vasopressin (AVP), the antidiuretic hormone, is a cyclic nonpeptide that is involved in the regulation of body fluid osmolality (1-3). AVP mediates its effects through a family of G-protein coupled receptors, the vasopressin receptors type V1a, V2 and V3 (also designated V1b) (1,2). The AVP receptor V1a is responsible for several functions, including blood vessel constriction, liver glycogenolysis and platelet adhesion (3). It is detected as a full length protein and a shorter protein, which results from proteolytic cleavage of its amino terminus (4). The V1a receptor is coupled to Gq/11 protein, which increases the intracellular calcium concentration (3). The human AVP receptor V2 gene maps to chromosome Xq28 and is expressed in lung and kidney (5,6). Mutations in the V2 receptor result in nephrogenic diabetes insipidus (NDI), a rare X-linked disorder characterized by the inability of the kidney to concentrate urine in response to AVP (5,7). The AVP Receptor V2 activates the Gs protein and the cyclic AMP second messenger system (7). The AVP receptor V3 is preferentially expressed in the pituitary and stimulates the release of adrenocorticotropic hormone (ACTH) in response to AVP by mobilizing intracellular calcium stores (8). AVP receptor antagonists may have potential therapeutic effects in hypertension, congestive heart failure, nephrotic syndrome and ACTH-secreting tumors (2).
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: AVPR1B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat