To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Amyotrophic lateral sclerosis 2 juvenile chromosome region candidate 1
- Antigen Symbol:ALS2CR1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:N3 NGG1 interacting factor 3-like 1|N3-like protein 1|N3 NGG1 interacting factor 3-like 1 S. pombe|S.pombe homolog-like 1|Ngg1 interacting factor 3-like 1|ALS2CR1|N3 Ngg1 interacting factor 3|Amyotrophic lateral sclerosis 2 juvenile chromosome region|N3L1.|candidate 1|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 1 protein|MDS015|N3L_HUMAN|CALS-7
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10260-580
- Supplier no.:BS-11649R-FITC
Specifications
About this item
The NGG1 interacting factor 3-like 1 (NIF3L1) is a 377 amino acid protein expressed mainly in the cytoplasm of cells in several different tissues. It has been highly conserved throughout evolution, from bacteria to mammals. NIF3L participates in retinoic acid-primed neural differentiation of P19 embryonic carcinoma cells by cooperating with Trip15/CSN2, a transcriptional corepressor/component of COP9 signalosome. NIF3L1 interacts with itself and with the NIF3L1 binding protein 1 (NIF3L1 BP1), which is a novel protein presumed to contain a leucine zipper domain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ALS2CR1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat