Specifications
- Antibody Type:Primary
- Antigen Name:SRY (sex determining region Y)-box 13
- Antigen Symbol:SOX13
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:MGC117216|SOX 13|ICA 12|sox13|SOX13 protein|Transcription factor Sox-13|Transcription factor SOX 13|ICA12|SRY related HMG box gene 13|Islet cell antigen 12|Type 1 diabetes autoantigen|SRY Sex determining region Y-box 13|SOX13_HUMAN|SRY box 13|SRY sex determining region Y box 13|SOX 13 protein|SRY box containing gene 13|Islet cell 12|Type 1 diabetes autoantigen ICA 12|Type 1 diabetes autoantigen ICA12.
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10262-740
- Supplier no.:BS-11586R-FITC
Specifications
About this item
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SOX13
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat